Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sts
Entrez Gene ID
24800
Gene Name
steroid sulfatase (microsomal), isozyme S
Gene Aliases
-
Chromosome Location
Chr.X: 45420469 - 45428748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012661.1 NP_036793.1 2 - 3 676 90
XM_006256960.2 XP_006257022.1 2 - 3 923 90
XM_006256961.2 XP_006257023.1 2 - 3 911 90
XM_006256962.2 XP_006257024.1 2 - 3 919 90
GenBank mRNA BC085747.1 - 1 - 2 161 90
U37138.1 - 2 - 3 676 90

View Full Product Details