The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012860.2 | NP_036992.2 | 3 - 4 | 457 | 63 |
GenBank mRNA | BC089770.1 | - | 3 - 4 | 457 | 63 |
X15734.1 | - | 3 - 4 | 364 | 63 | |
X60822.1 | - | 3 - 4 | 543 | 63 |