The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013008.3 | NP_037140.2 | 5 - 6 | 786 | 66 |
XM_006247974.1 | XP_006248036.1 | 5 - 6 | 1070 | 66 | |
GenBank mRNA | AB622208.1 | - | 4 - 5 | 797 | 66 |
BC061563.1 | - | 5 - 6 | 790 | 66 | |
BC061564.1 | - | 5 - 6 | 805 | 66 | |
L01506.1 | - | 5 - 6 | 754 | 66 | |
L01507.1 | - | 5 - 6 | 830 | 66 | |
M23253.1 | - | 5 - 6 | 700 | 66 | |
X12658.1 | - | 5 - 6 | 723 | 66 |