Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hnf1b
Entrez Gene ID
25640
Gene Name
HNF1 homeobox B
Gene Aliases
LF-B3, Tcf2, VHNF1
Chromosome Location
Chr.10: 71159984 - 71218607 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013103.1 NP_037235.1 8 - 9 1561 81
GenBank mRNA BC081826.1 - 7 - 8 1676 81
EF121973.1 - 8 - 9 1534 81
EF121974.1 - 8 - 9 1456 81
EF122006.1 - 8 - 9 1534 81
X56546.1 - 8 - 9 1561 81

View Full Product Details