Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Akt2
Entrez Gene ID
25233
Gene Name
v-akt murine thymoma viral oncogene homolog 2
Gene Aliases
-
Chromosome Location
Chr.1: 84400940 - 84451223 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017093.1 NP_058789.1 5 - 6 725 69
XM_008759108.1 XP_008757330.1 6 - 7 3124 69
XM_008759109.1 XP_008757331.1 5 - 6 712 69
XM_008759110.1 XP_008757332.1 5 - 6 2989 69
XM_008759111.1 XP_008757333.1 5 - 6 835 69
GenBank mRNA D30041.1 - 5 - 6 725 69

View Full Product Details