Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tceb3
Entrez Gene ID
25562
Gene Name
transcription elongation factor B (SIII), polypeptide 3
Gene Aliases
El
Chromosome Location
Chr.5: 154348167 - 154363324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017103.2 NP_058799.2 5 - 6 1597 75
GenBank mRNA BC081815.1 - 5 - 6 1597 75
L46816.1 - - 1612 75

View Full Product Details