Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Drd3
Entrez Gene ID
29238
Gene Name
dopamine receptor D3
Gene Aliases
-
Chromosome Location
Chr.11: 61822077 - 61883223 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017140.1 NP_058836.1 4 - 5 805 56
XM_006248313.2 XP_006248375.2 5 - 6 1022 56
GenBank mRNA X53944.1 - 4 - 5 805 56

View Full Product Details