The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017176.2 | NP_058872.1 | 3 - 4 | 359 | 68 |
GenBank mRNA | AF166100.1 | - | 3 - 4 | 361 | 68 |
AY651029.1 | - | 3 - 4 | 355 | 68 |