Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vipr2
Entrez Gene ID
29555
Gene Name
vasoactive intestinal peptide receptor 2
Gene Aliases
-
Chromosome Location
Chr.6: 143937961 - 144007028 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017238.1 NP_058934.1 5 - 6 565 95
GenBank mRNA U09631.1 - 5 - 6 565 95
Z25885.1 - 5 - 6 518 95

View Full Product Details