Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc24a1
Entrez Gene ID
56814
Gene Name
solute carrier family 24 (sodium/potassium/calcium exchanger), member 1
Gene Aliases
Nckx1
Chromosome Location
Chr.8: 70411186 - 70436337 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020090.1 NP_064475.1 6 - 7 3043 70
GenBank mRNA AF176688.1 - 6 - 7 3043 70

View Full Product Details