Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acvrl1
Entrez Gene ID
25237
Gene Name
activin A receptor type II-like 1
Gene Aliases
R-3, R3, SETHKIR
Chromosome Location
Chr.7: 142770486 - 142787336 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022441.2 NP_071886.1 7 - 8 1194 76
XM_006242303.2 XP_006242365.1 8 - 9 1595 76
XM_006242304.2 XP_006242366.1 8 - 9 2199 76
XM_006242305.2 XP_006242367.1 7 - 8 2214 76
XM_006242307.2 XP_006242369.1 7 - 8 2084 76
XM_008765673.1 XP_008763895.1 8 - 9 2258 76
GenBank mRNA BC083173.1 - 7 - 8 1194 76
FQ223580.1 - 7 - 8 1236 76
L36088.1 - 8 - 9 1609 76

View Full Product Details