The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031093.2 | NP_112355.1 | 3 - 4 | 531 | 79 |
XM_006253993.2 | XP_006254055.1 | 4 - 5 | 735 | 79 | |
XM_008771763.1 | XP_008769985.1 | 4 - 5 | 549 | 79 |