Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnma1
Entrez Gene ID
83731
Gene Name
potassium channel, calcium activated large conductance subfamily M alpha, member 1
Gene Aliases
BKCa, KCNMA1b, KCNMA1c, Kcnma, Slo
Chromosome Location
Chr.15: 344204 - 1048849 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031828.1 NP_114016.1 14 - 15 2248 124
GenBank mRNA AF083341.1 - 14 - 15 1684 124
AF135265.1 - 14 - 15 2102 124
AY330290.1 - 14 - 15 1573 124
AY330291.1 - 14 - 15 1573 124
AY330292.1 - 14 - 15 1582 124
AY330293.2 - 14 - 15 1573 124
AY330294.1 - 14 - 15 1573 124
AY344964.1 - 14 - 15 1573 124
AY344965.2 - 13 - 14 1985 124
U40603.1 - 14 - 15 2248 124
U93052.1 - 14 - 15 1674 124

View Full Product Details