Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnma1
Entrez Gene ID
83731
Gene Name
potassium channel, calcium activated large conductance subfamily M alpha, member 1
Gene Aliases
BKCa, KCNMA1b, KCNMA1c, Kcnma, Slo
Chromosome Location
Chr.15: 344204 - 1048849 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031828.1 NP_114016.1 23 - 24 2999 61
GenBank mRNA AF083341.1 - 21 - 22 2252 61
AF135265.1 - 21 - 22 2670 61
AY330291.1 - 20 - 21 2129 61
AY330292.1 - 20 - 21 2138 61
AY330293.2 - 20 - 21 2129 61
AY344964.1 - 18 - 19 1965 61
AY344965.2 - 19 - 20 2541 61
U40603.1 - 23 - 24 2999 61
U55995.1 - 20 - 21 2282 61
U93052.1 - 20 - 21 2230 61

View Full Product Details