Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clec4f
Entrez Gene ID
114598
Gene Name
C-type lectin domain family 4, member F
Gene Aliases
Clecsf13, Kclr
Chromosome Location
Chr.4: 115322300 - 115332021 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053753.1 NP_446205.1 6 - 7 1535 73
GenBank mRNA BC098661.1 - 6 - 7 1566 73
J03734.1 - 6 - 7 1526 73

View Full Product Details