Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpina12
Entrez Gene ID
191570
Gene Name
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12
Gene Aliases
Ol-64, Ol64
Chromosome Location
Chr.6: 127696501 - 127711238 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138825.2 NP_620180.2 2 - 3 846 84
GenBank mRNA AF245398.1 - 2 - 3 846 84

View Full Product Details