The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012928.1 | NP_037060.1 | - | 935 | 108 |
GenBank mRNA | AF126808.1 | - | - | 829 | 108 |
X55519.1 | - | - | 935 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008759081.1 | XP_008757303.1 | 2 - 3 | 287 | 108 |