Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcns3
Entrez Gene ID
83588
Gene Name
potassium voltage-gated channel, modifier subfamily S, member 3
Gene Aliases
-
Chromosome Location
Chr.6: 36764040 - 36819810 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031778.2 NP_113966.2 2 - 3 307 62
GenBank mRNA Y17607.1 - 2 - 3 307 62

View Full Product Details