Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kcnk4
Entrez Gene ID
116489
Gene Name
potassium channel, two pore domain subfamily K, member 4
Gene Aliases
KT4.1, TRAAK
Chromosome Location
Chr.1: 222182997 - 222192039 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053804.2 NP_446256.2 3 - 4 474 54
XM_006230634.2 XP_006230696.1 4 - 5 1772 54
XM_008760054.1 XP_008758276.1 4 - 5 1515 54
XM_008760055.1 XP_008758277.1 4 - 5 1386 54
XM_008760056.1 XP_008758278.1 4 - 5 592 54
GenBank mRNA AF302842.2 - 3 - 4 474 54

View Full Product Details