Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a24
Entrez Gene ID
286961
Gene Name
solute carrier family 22, member 24
Gene Aliases
Oat5, Slc22a19, Slc22a9
Chromosome Location
Chr.1: 222969899 - 222982141 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173302.1 NP_775424.1 - 525 115
GenBank mRNA AB051836.1 - - 525 115

View Full Product Details