The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013143.1 | NP_037275.1 | 8 - 9 | 816 | 70 |
XM_008766837.1 | XP_008765059.1 | 8 - 9 | 855 | 70 | |
XM_008766838.1 | XP_008765060.1 | 6 - 7 | 677 | 70 | |
GenBank mRNA | BC081834.1 | - | 8 - 9 | 831 | 70 |
S43408.1 | - | 8 - 9 | 816 | 70 |