The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181366.2 | NP_852031.1 | 3 - 4 | 204 | 97 |
GenBank mRNA | AF538953.1 | - | 3 - 4 | 214 | 97 |