The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181385.2 | NP_852050.1 | 3 - 4 | 967 | 61 |
GenBank mRNA | AY283175.1 | - | 3 - 4 | 967 | 61 |