Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Grpcb
- Entrez Gene ID
-
360395
- Gene Name
- glutamine/glutamic acid-rich protein A
- Gene Aliases
-
CRP, GRP-CB
- Chromosome Location
-
Chr.4: 167481724 - 167587767 on Build Rnor_6.0
- Gene Symbol
- Grpca
- Entrez Gene ID
-
192266
- Gene Name
- glutamine/glutamic acid-rich protein A
- Gene Aliases
-
Grp-Ca, Grpca_m
- Chromosome Location
-
-
- Gene Symbol
- LOC100360165
- Entrez Gene ID
-
100360165
- Gene Name
- rCG29588-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 167419917 - 167423526 on Build Rnor_6.0
View Full Product Details