The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181639.3 | NP_853670.2 | 4 - 5 | 686 | 111 |
GenBank mRNA | AY273196.1 | - | 3 - 4 | 608 | 111 |
BC078678.1 | - | 4 - 5 | 686 | 111 |