Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsb66
Entrez Gene ID
296610
Gene Name
Rsb-66 protein
Gene Aliases
-
Chromosome Location
Chr.3: 7269851 - 7279356 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181694.2 NP_859045.2 4 - 5 584 63
XM_006233766.1 XP_006233828.1 4 - 5 747 63
GenBank mRNA AY121839.1 - 3 - 4 534 63
BC097268.1 - 4 - 5 584 63

View Full Product Details