The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182669.2 | NP_872610.2 | 7 - 8 | 1853 | 80 |
GenBank mRNA | AY149342.1 | - | 4 - 5 | 1293 | 80 |
BC098647.1 | - | 7 - 8 | 1853 | 80 |