Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSA-14-44
Entrez Gene ID
297173
Gene Name
RSA-14-44 protein
Gene Aliases
-
Chromosome Location
Chr.4: 96831880 - 96833252 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182670.2 NP_872611.2 1 - 1 964 113
GenBank mRNA AY149343.1 - 1 - 1 921 113
BC083656.1 - 1 - 1 953 113

View Full Product Details