The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077826.1 | NP_001071294.1 | 2 - 3 | 243 | 59 |
GenBank mRNA | BC158557.1 | - | - | 346 | 59 |
EF044301.1 | - | 2 - 3 | 243 | 59 | |
FQ213412.1 | - | 2 - 3 | 254 | 59 |