The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022385.3 | NP_071780.1 | 4 - 5 | 426 | 73 |
GenBank mRNA | BC061553.1 | - | - | 509 | 73 |
FQ226660.1 | - | 4 - 5 | 482 | 73 | |
U12402.1 | - | 4 - 5 | 358 | 73 | |
X76920.1 | - | 4 - 5 | 426 | 73 |