The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031354.1 | NP_112644.1 | 6 - 7 | 750 | 131 |
XM_006251661.1 | XP_006251723.1 | 7 - 8 | 871 | 131 | |
GenBank mRNA | AB039663.1 | - | 6 - 7 | 750 | 131 |
AF268468.1 | - | 5 - 6 | 586 | 131 | |
BC063164.1 | - | 6 - 7 | 719 | 131 | |
FQ217146.1 | - | 3 - 4 | 475 | 131 | |
FQ219952.1 | - | 3 - 4 | 475 | 130 | |
FQ228796.1 | - | 3 - 4 | 475 | 131 |