The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004271.1 | NP_001004271.1 | 1 - 2 | 743 | 151 |
XM_008770060.1 | XP_008768282.1 | 1 - 2 | 770 | 151 | |
GenBank mRNA | BC078782.1 | - | 1 - 2 | 743 | 151 |
FQ219334.1 | - | 1 - 2 | 237 | 151 | |
U06273.1 | - | 1 - 2 | 724 | 151 | |
U06274.1 | - | 1 - 2 | 725 | 151 |