Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prl3d4
Entrez Gene ID
24282
Gene Name
prolactin family 3, subfamily d, member 4
Gene Aliases
Csh1l1, Csh1v, Pl-Iv
Chromosome Location
Chr.17: 39271883 - 39278716 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033233.1 NP_150236.1 4 - 5 498 137
XM_008771595.1 XP_008769817.1 4 - 5 480 137
GenBank mRNA BC097302.1 - 4 - 5 524 137
U32679.1 - 4 - 5 498 137

Gene Symbol
LOC103689989
Entrez Gene ID
103689989
Gene Name
prolactin-3D4
Gene Aliases
-
Chromosome Location
Chr.17: 42920887 - 42927745 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771673.1 XP_008769895.1 - 480 137
XM_008771674.1 XP_008769896.1 - 518 137

View Full Product Details