The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130755.1 | NP_570111.1 | 7 - 8 | 859 | 71 |
GenBank mRNA | AF461496.1 | - | 7 - 8 | 859 | 71 |
DQ403126.1 | - | 1 - 2 | 122 | 71 |