The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182843.1 | NP_878263.1 | 7 - 8 | 1427 | 85 |
GenBank mRNA | AY336075.1 | - | 7 - 8 | 1427 | 85 |
BC083877.1 | - | 7 - 8 | 1388 | 85 |