Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chi3l1
Entrez Gene ID
89824
Gene Name
chitinase 3-like 1 (cartilage glycoprotein-39)
Gene Aliases
Chil1
Chromosome Location
Chr.13: 51022826 - 51030797 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006249892.2 XP_006249954.2 1 - 2 137 63

View Full Product Details