The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017186.2 | NP_058882.1 | 5 - 6 | 733 | 120 |
GenBank mRNA | AF081557.1 | - | 5 - 6 | 734 | 120 |