The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017285.1 | NP_058981.1 | 5 - 6 | 636 | 102 |
GenBank mRNA | BC084723.1 | - | 4 - 5 | 559 | 102 |
D21800.1 | - | 5 - 6 | 636 | 102 | |
FQ211229.1 | - | 5 - 6 | 593 | 102 | |
FQ232780.1 | - | 5 - 6 | 632 | 102 |