The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019356.1 | NP_062229.1 | 6 - 7 | 703 | 92 |
GenBank mRNA | BC087019.1 | - | 6 - 7 | 760 | 92 |
J02646.1 | - | 6 - 7 | 703 | 92 |