Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cox7a2
Entrez Gene ID
29507
Gene Name
cytochrome c oxidase subunit VIIa polypeptide 2
Gene Aliases
Cox7a3
Chromosome Location
Chr.8: 87209529 - 87213627 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022503.2 NP_071948.1 2 - 3 192 94
GenBank mRNA AW521400.1 - 2 - 3 94
BC166880.1 - 2 - 3 160 94
BG668415.1 - 2 - 3 199 94
FQ209801.1 - 2 - 3 187 94
FQ209847.1 - 2 - 3 159 94
FQ210551.1 - 2 - 3 159 94
FQ211276.1 - 2 - 3 160 94
FQ214317.1 - 2 - 3 168 94
FQ217361.1 - 2 - 3 159 94
FQ221839.1 - 2 - 3 164 94
FQ223968.1 - 2 - 3 183 94
FQ228537.1 - 2 - 3 231 94
FQ229029.1 - 2 - 3 161 94
FQ229035.1 - 2 - 3 193 94
FQ230547.1 - 2 - 3 220 94
X54080.1 - 2 - 3 166 94

Gene Symbol
Cox7a2l2
Entrez Gene ID
688386
Gene Name
cytochrome c oxidase subunit VIIa polypeptide 2-like 2
Gene Aliases
-
Chromosome Location
Chr.14: 52661933 - 52662375 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751367.3 XP_003751415.1 1 - 1 143 94

View Full Product Details