The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024139.2 | NP_077053.1 | 5 - 6 | 544 | 107 |
GenBank mRNA | AB070350.1 | - | 5 - 6 | 529 | 107 |
BC062029.1 | - | 5 - 6 | 544 | 107 | |
U39875.1 | - | 5 - 6 | 790 | 107 |