The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024351.2 | NP_077327.1 | 8 - 9 | 1835 | 63 |
GenBank mRNA | BC061547.1 | - | 8 - 9 | 1842 | 63 |
BC098914.1 | - | 8 - 9 | 1787 | 63 | |
FQ213866.1 | - | 4 - 5 | 851 | 63 | |
FQ221758.1 | - | 1 - 2 | 70 | 63 | |
FQ226406.1 | - | 4 - 5 | 770 | 63 | |
FQ231618.1 | - | 3 - 4 | 482 | 63 | |
FQ231904.1 | - | 4 - 5 | 1027 | 63 | |
FQ232261.1 | - | 4 - 5 | 770 | 63 | |
FQ235115.1 | - | 4 - 5 | 1184 | 63 | |
M11942.1 | - | 8 - 9 | 1806 | 63 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008764620.1 | XP_008762842.1 | 1 - 1 | 1837 | 63 |