The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031104.1 | NP_112366.1 | 4 - 5 | 260 | 98 |
GenBank mRNA | BC058466.1 | - | 3 - 4 | 276 | 98 |
BC082750.1 | - | 3 - 4 | 251 | 98 | |
FQ222147.1 | - | 3 - 4 | 269 | 98 | |
FQ222660.1 | - | 3 - 4 | 269 | 98 | |
FQ224693.1 | - | 3 - 4 | 268 | 98 | |
FQ228373.1 | - | 3 - 4 | 266 | 98 | |
X78444.1 | - | 4 - 5 | 260 | 98 |