Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmed10
Entrez Gene ID
84599
Gene Name
transmembrane emp24-like trafficking protein 10 (yeast)
Gene Aliases
Tmp21
Chromosome Location
Chr.6: 109169739 - 109205004 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053467.1 NP_445919.1 4 - 5 542 81
GenBank mRNA AJ004912.1 - 4 - 5 542 81
FQ212056.1 - 4 - 5 576 81
X97443.1 - 4 - 5 494 81

View Full Product Details