Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp5j
Entrez Gene ID
94271
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F6
Gene Aliases
-
Chromosome Location
Chr.11: 24286806 - 24294858 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053602.2 NP_446054.1 2 - 3 325 124
XM_008768600.1 XP_008766822.1 3 - 4 725 124
XM_008768601.1 XP_008766823.1 3 - 4 449 124
XM_008768602.1 XP_008766824.1 3 - 4 399 124
XM_008768603.1 XP_008766825.1 2 - 3 299 124
GenBank mRNA BC059121.1 - 2 - 3 124
FQ212150.1 - 2 - 3 228 124
FQ214305.1 - 2 - 3 228 124
FQ216982.1 - 2 - 3 220 124
FQ217271.1 - 2 - 3 226 124
FQ217448.1 - 2 - 3 232 124
FQ224472.1 - 2 - 3 228 124
FQ228382.1 - 2 - 3 228 124
M73030.1 - 2 - 3 212 124
X54510.1 - 2 - 3 325 124

View Full Product Details