Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dpp3
Entrez Gene ID
114591
Gene Name
dipeptidylpeptidase 3
Gene Aliases
-
Chromosome Location
Chr.1: 220166124 - 220189676 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053748.1 NP_446200.1 11 - 12 1397 123
XM_006230633.2 XP_006230695.1 12 - 13 1584 123
GenBank mRNA BC107673.1 - 12 - 13 1408 123
D89340.2 - 11 - 12 1397 123

View Full Product Details