Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fcgr2a
- Entrez Gene ID
-
116591
- Gene Name
- Fc fragment of IgG, low affinity IIa, receptor
- Gene Aliases
-
Fcgr3, Fcgr3a
- Chromosome Location
-
Chr.13: 91146879 - 91163694 on Build Rnor_6.0
- Gene Symbol
- LOC103693683
- Entrez Gene ID
-
103693683
- Gene Name
- low affinity immunoglobulin gamma Fc region receptor III
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 89420835 - 89440644 on Build Rnor_6.0
View Full Product Details