The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053982.1 | NP_446434.1 | 3 - 4 | 308 | 68 |
XM_006230097.1 | XP_006230159.1 | 4 - 5 | 355 | 68 | |
XM_006230098.1 | XP_006230160.1 | 4 - 5 | 398 | 68 | |
GenBank mRNA | BC058452.1 | - | 4 - 5 | 333 | 68 |
FQ210209.1 | - | 4 - 5 | 330 | 68 | |
FQ212042.1 | - | 4 - 5 | 329 | 68 | |
FQ212257.1 | - | 4 - 5 | 327 | 68 | |
FQ214481.1 | - | 4 - 5 | 329 | 68 | |
FQ220927.1 | - | 4 - 5 | 325 | 68 | |
FQ220997.1 | - | 4 - 5 | 329 | 68 | |
FQ221041.1 | - | 4 - 5 | 329 | 68 | |
FQ221174.1 | - | 4 - 5 | 327 | 68 | |
FQ221230.1 | - | 4 - 5 | 402 | 68 | |
FQ221308.1 | - | 4 - 5 | 331 | 68 | |
FQ221419.1 | - | 4 - 5 | 329 | 68 | |
FQ221518.1 | - | 4 - 5 | 329 | 68 | |
FQ221677.1 | - | 4 - 5 | 327 | 68 | |
FQ221720.1 | - | 4 - 5 | 398 | 68 | |
FQ221825.1 | - | 4 - 5 | 329 | 68 | |
FQ221840.1 | - | 4 - 5 | 329 | 68 | |
FQ221877.1 | - | 4 - 5 | 329 | 68 | |
FQ222252.1 | - | 4 - 5 | 340 | 68 | |
FQ222272.1 | - | 4 - 5 | 329 | 68 | |
FQ222274.1 | - | 4 - 5 | 329 | 68 | |
FQ222304.1 | - | 4 - 5 | 329 | 68 | |
FQ222376.1 | - | 4 - 5 | 329 | 68 | |
FQ222415.1 | - | 4 - 5 | 327 | 68 | |
FQ222418.1 | - | 4 - 5 | 331 | 68 | |
FQ222718.1 | - | 4 - 5 | 329 | 68 | |
FQ222721.1 | - | 4 - 5 | 327 | 68 | |
FQ222768.1 | - | 4 - 5 | 327 | 68 | |
FQ222892.1 | - | 4 - 5 | 330 | 68 | |
FQ222921.1 | - | 4 - 5 | 332 | 68 | |
FQ222922.1 | - | 4 - 5 | 398 | 68 | |
FQ223006.1 | - | 4 - 5 | 329 | 68 | |
FQ223095.1 | - | 4 - 5 | 329 | 68 | |
FQ223230.1 | - | 4 - 5 | 328 | 68 | |
FQ223341.1 | - | 4 - 5 | 329 | 68 | |
FQ223389.1 | - | 4 - 5 | 329 | 68 | |
FQ223433.1 | - | 4 - 5 | 329 | 68 | |
FQ223787.1 | - | 4 - 5 | 329 | 68 | |
FQ224201.1 | - | 4 - 5 | 329 | 68 | |
FQ224347.1 | - | 4 - 5 | 329 | 68 | |
FQ224383.1 | - | 4 - 5 | 329 | 68 | |
FQ228419.1 | - | 4 - 5 | 328 | 68 | |
FQ228429.1 | - | 4 - 5 | 329 | 68 | |
FQ228446.1 | - | 4 - 5 | 329 | 68 | |
FQ228475.1 | - | 4 - 5 | 330 | 68 | |
FQ228508.1 | - | 4 - 5 | 329 | 68 | |
FQ228609.1 | - | 4 - 5 | 329 | 68 | |
FQ228617.1 | - | 4 - 5 | 328 | 68 | |
FQ229042.1 | - | 4 - 5 | 327 | 68 | |
FQ229063.1 | - | 4 - 5 | 328 | 68 | |
X77953.1 | - | 3 - 4 | 308 | 68 |