Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmb6
Entrez Gene ID
29666
Gene Name
proteasome (prosome, macropain) subunit, beta type 6
Gene Aliases
Psmb6l
Chromosome Location
Chr.10: 57131347 - 57133685 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057099.3 NP_476440.2 4 - 5 432 124
XM_008767811.1 XP_008766033.1 4 - 5 463 124
GenBank mRNA BC058451.1 - 4 - 5 436 124
D10754.1 - 4 - 5 421 124
FQ209811.1 - 4 - 5 435 124
FQ217385.1 - 4 - 5 435 124
FQ224639.1 - 4 - 5 435 124

Gene Symbol
LOC100360846
Entrez Gene ID
100360846
Gene Name
proteasome subunit beta type 6-like
Gene Aliases
-
Chromosome Location
Chr.2: 96031948 - 96033565 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729028.3 XP_002729074.2 2 - 2 580 124

View Full Product Details