The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134398.2 | NP_599225.1 | 3 - 4 | 396 | 72 |
XM_006243223.2 | XP_006243285.1 | 3 - 4 | 428 | 72 | |
GenBank mRNA | AF178669.1 | - | 3 - 4 | 398 | 72 |
FQ224819.1 | - | 3 - 4 | 371 | 72 |